missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554123 Supplier Invitrogen™ Supplier No. PA554123
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: AVQRRSRLQR RQSFAVLRGA VLGLQDGGDN DDAAEASSEP TEKAPSEEEL HGDQTDFGQG SQSPQKQEEQ RQHLHLMVQL LRPQDDIRLA AQLEAPRPPR LRY Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.

The ADF (actin-depolymerizing factor)/cofilin family is composed of stimulus-responsive mediators of actin dynamics. ADF/cofilin proteins are inactivated by kinases such as LIM domain kinase-1 (LIMK1). The SSH family appears to play a role in actin dynamics by reactivating ADF/cofilin proteins in vivo.

Specifications

Antigen SSH3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ssh3
Gene Accession No. Q8TE77
Gene Alias BC028922; hSSH-3L; mSSH-3L; Protein phosphatase Slingshot homolog 3; slingshot 3; slingshot homolog 3; slingshot homolog 3 (Drosophila); slingshot protein phosphatase 3; slingshot-3L; slingshot-like 3; Ssh3; SSH-3; Ssh3l; SSH-3L; SSH-like protein 3
Gene Symbols Ssh3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SSH3. Recombinant protein control fragment (Product #RP-91578).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54961
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less