missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSEA3 Monoclonal Antibody (MC-631)

Rat Monoclonal Antibody

$315.71 - $721.24

Specifications

Antigen SSEA3
Clone MC-631
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications Flow Cytometry, Immunocytochemistry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PIMA1020
View Documents
Invitrogen™
MA1020
100 μg
Each for $721.24
Only null left
 
PIMA1020X
View Documents
Invitrogen™
MA1020X
20 μL
Each for $315.71
Only null left
 
Description

Description

SSEA3 Monoclonal Antibody for Flow, ICC/IF

SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications

Specifications

SSEA3
1 mg/mL
Flow Cytometry, Immunocytochemistry
Unconjugated
Rat
RUO
Potassium phosphate with 0.15M NaCl, 1mg/mL BSA and 0.05% sodium azide
Fischer rat immunized with 4-8 cell stage mouse embryos.
Antibody
MC-631
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgM
Human
SSEA-3; stage-specific embryonic antigen 3
Primary
Affinity chromatography - MBP
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'