missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSEA1 Recombinant Mouse Monoclonal Antibody (BRA-4F1)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551828
Description
Specificity: This antibody is specific for CD15. It also shows crossreactivity with the CD15 antigen from other species.
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.Specifications
| SSEA1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P22083 | |
| FUT4 | |
| This antibody was raised by immunzing mice with human CD15. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| BRA-4F1 | |
| Unconjugated | |
| FUT4 | |
| FUTIV; Lewis X; SSEA-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |