missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SSC buffer (20X)

Catalog No. AAJ60839K2
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60839K2 1 L
AAJ60839K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60839K2 Supplier Thermo Scientific Chemicals Supplier No. J60839K2

Suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

SSC buffer is suitable for preparing nucleic acids for hybridization, hybridization buffer for northern and southern blots, washing buffer for northern and southern blots. Saline-Sodium Citrate Buffer (SSC) is used as a hybridization buffer in immunoblotting and DNA Microarray protocols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Liquid
Quantity 1 L

RUO – Research Use Only