missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRPK2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5149023
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection. It therefore represents an important host cell target for therapeutic intervention in HBV infection. Promotes neuronal apoptosis by up-regulating cyclin-D1 (CCND1) expression. This is done by the phosphorylation of SRSF2, leading to the suppression of p53/TP53 phosphorylation thereby relieving the repressive effect of p53/TP53 on cyclin-D1 (CCND1) expression. Phosphorylates ACIN1, and redistributes it from the nuclear speckles to the nucleoplasm, resulting in cyclin A1 but not cyclin A2 up-regulation. Plays an essential role in spliceosomal B complex formation via the phosphorylation of DDX23/PRP28. Plays a negative role in the regulation of HBV replication through a mechanism not involving the phosphorylation of the core protein but by reducing the packaging efficiency of the pregenomic RNA (pgRNA) without affecting the formation of the viral core particles .Chemical Identifiers
| SRPK2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human | |
| SRPK2 | |
| AW226533; AW492537; AW547358; H_RG152G17.1a; H_RG152G17.1b; mSRPK2; serine kinase SRPK2; serine/arginine-rich protein specific kinase 2; serine/arginine-rich protein-specific kinase 2; serine/arginine-rich splicing factor kinase 2; serine/threonine-protein kinase SRPK2; SFRS protein kinase 2; SFRSK2; SR protein kinase 2; SRPK2; SR-protein-specific kinase 2; SRSF protein kinase 2; SRSF protein kinase 2 C-terminal; SRSF protein kinase 2 N-terminal; Wbp6; WUGSC:H_RG152G17.1a; WW domain binding protein 6 | |
| SRPK2 | |
| Primary | |
| Antigen affinity chromatography |
| 0.2 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Goat | |
| RUO | |
| PBS with trehalose and no preservative | |
| P78362 | |
| 6733 | |
| E. coli-derived recombinant human SRPK2, Asn313-Ala466. | |
| Antibody |
Specifications
| SRPK2 | |
| Polyclonal | |
| Unconjugated | |
| SRPK2 | |
| AW226533; AW492537; AW547358; H_RG152G17.1a; H_RG152G17.1b; mSRPK2; serine kinase SRPK2; serine/arginine-rich protein specific kinase 2; serine/arginine-rich protein-specific kinase 2; serine/arginine-rich splicing factor kinase 2; serine/threonine-protein kinase SRPK2; SFRS protein kinase 2; SFRSK2; SR protein kinase 2; SRPK2; SR-protein-specific kinase 2; SRSF protein kinase 2; SRSF protein kinase 2 C-terminal; SRSF protein kinase 2 N-terminal; Wbp6; WUGSC:H_RG152G17.1a; WW domain binding protein 6 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 6733 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P78362 | |
| SRPK2 | |
| E. coli-derived recombinant human SRPK2, Asn313-Ala466. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |