Learn More
Invitrogen™ Human SRMS (SRM), GST Tag Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ PV4214
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
SRMS (Srm) is a tyrosine protein kinase (SRC sub-family) that may be involved in the proliferation and differentiation of keratinocytes. Non-receptor tyrosine-protein kinase which phosphorylates DOK1 on tyrosine residues. Also phosphorylates KHDRBS1/SAM68 and VIM on tyrosine residues. Phosphorylation of KHDRBS1 is EGF-dependent.Specifications
| Q9H3Y6 | |
| Kinase Assay | |
| 6725 | |
| Human SRMS (SRM), GST Tag | |
| For research use only. Not for use in diagnostic procedures | |
| SRMS | |
| SRMS (SRM) Recombinant Human Protein | |
| Human | |
| GST-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Kinase and inhibitors | |
| Cytoplasmic tyrosine kinase |
| See Label | |
| 50 mM HEPES with 0.02% Triton X-100, 0.5 mM EDTA, 2 mM DTT, 50% glycerol and no preservative; pH 7 | |
| 82 kDa | |
| 10 μg | |
| A230069J08Rik; C20orf148; dJ697K14.1; PTK70; src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites; SRM; SRMS; Tyrosine-protein kinase Srms | |
| SRC | |
| Unconjugated | |
| Recombinant | |
| Full length | |
| Insect cells | |
| Liquid |