missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. 720240
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
720240 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 720240 Supplier Invitrogen™ Supplier No. LS720240
Only null left

Rabbit Polyclonal Antibody

These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Bovine and Pig.

Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.

Specifications

Antigen SRF
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2
Gene SRF
Gene Accession No. P11831, Q9JM73
Gene Alias AW049942; AW240594; c-fos serum response element-binding transcription factor; MCM1; minichromosome maintenance 1 homolog; OTTHUMP00000039820; RGD1559787; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); Srf
Gene Symbols SRF
Host Species Rabbit
Immunogen Peptides corresponding to Human SRF (aa 223-238,248-263,370-385).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20807, 501099, 6722
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less