missing translation for 'onlineSavingsMsg'
Learn More

SREBP-1, Mouse anti-Hamster, Canine, Human, Clone: 2A4, Millipore Sigma™

Catalog No. p-7162966
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS203425 25 μg
MABS2034100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS203425 Supplier MilliporeSigma Supplier No. MABS203425UG
Only null left

Mouse Monoclonal Antibody

Anti-SREBP-1, clone 2A4, Cat. No. MABS2034, is a mouse monoclonal antibody that detects Sterol regulatory element-binding protein 1 and has been tested for use in Western Blotting.

Sterol regulatory element-binding protein 1 (UniProt: P36956; also known as SREBP-1, Class D basic helix-loop-helix protein 1, bHLHd1, Sterol regulatory element-binding transcription factor 1) is encoded by the SREBF1 (also known as BHLHD1, SREBP1) gene (Gene ID: 6720) in human. SREBP-1 is a multi-pass membrane protein found on nuclear, endoplasmic reticulum, and Golgi membranes. It attaches to membranes via two hydrophobic sequences (aa 477-497 and 536-556). It is expressed in a wide variety of tissues, but is most abundant in liver and adrenal gland. Six isoforms of SREBP-1 have been described that are produced by alternative splicing. Isoform SREBP-1C predominates in liver, adrenal gland, and ovary, whereas isoform SREBP-1A predominates in hepatoma cell lines. Both isoforms are found in kidney, brain, white fat, and muscle. It serves as a transcriptional activator that is essential for lipid homeostasis and regulates transcription of the LDL receptor gene. It is also involved in fatty acid and cholesterol synthesis. It acts by binds to the sterol regulatory element 1 (SRE-1) (5'-ATCACCCCAC-3') and displays dual sequence specificity binding to both an E-box motif (5'-ATCACGTGA-3') and to SRE-1 (5'-ATCACCCCAC-3'). SREBP-1 has two cytoplasmic domains (aa 1-487 and 569-1147), two transmembrane domains (aa 88-508 and 548-568), and one lumenal domain (aa 509-547). In the precursor form, which has a hairpin orientation in the membrane both the N-terminal transcription factor domain and the C-terminal regulatory domain face the cytoplasm. Two separate site-specific proteolytic cleavages are required for release of the transcriptionally active N-terminal domain. These cleavages are carried out by two distinct proteases, called site-1 protease (S1P; aa 490-491) and site-2 protease (S2P; aa 530-531). SREBP-1 can be phosphorylated by AMPK at Ser402, which suppress its processing and nuclear translocation, and thereby repress target gene expression.

Specifications

Antigen SREBP-1
Applications Western Blot
Classification Monoclonal
Clone 2A4
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected SREBP-1 in Western Blotting applications (Toth, J.I., et. al. (2004). Mol Cell Biol. 24(18):8288-300; Key, C.C., et. al. (2017). Cell Rep. 19(10):2116-2129; Rosenfeld, J.M., et. al. (1998). J Biol Chem. 273(26):16112-21; Sato, R., et. al. (1994). J Biol Chem. 269(25):17267-73).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Sterol regulatory element-binding protein 1;Class D basic helix-loop-helix protein 1;bHLHd1;Sterol regulatory element-binding transcription factor 1
Gene Symbols SREBF1;BHLHD1;SREBP1
Host Species Mouse
Immunogen His-tagged recombinant fragment corresponding to 107 amino acids from the N-terminal half of human Sterol regulatory element-binding protein 1 (SREBP-1).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001005291.1
Test Specificity Clone 2A4 is a mouse monoclonal antibody that detects Sterol regulatory element-binding protein 1. It targets an epitope within 107 amino acids from the N-terminal half.
Target Species Hamster, Canine, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less