missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRC1 Monoclonal Antibody (1135/H4)
GREENER_CHOICE

Catalog No. MA1840
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1840 Supplier Invitrogen™ Supplier No. MA1840
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

MA1-840 detects steroid receptor coactivator-1 (SRC-1) from mouse and primate tissues and cells as well as recombinant human SRC-1 protein. MA1-840 has been successfully used in Western blot and immunoprecipitation procedures. By Western blot MA1-840 detects an ~165 kDa protein representing SRC-1 in COS cell extract. Also by Western blot, MA1-840 detects an unidentified protein at ~30-40 kDa. This antibody does not react with NIH-3T3 cells in Western blot. The MA1-840 immunogen is a synthetic peptide corresponding to residues 477-947 of human SRC-1. The epitope for MA1-840 has been mapped to the extreme C-terminal end of this fragment.

Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.

Specifications

Antigen SRC1
Applications ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1135/H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene NCOA1
Gene Accession No. P70365, Q15788
Gene Alias ASV; bHLHe42; bHLHe74; class E basic helix-loop-helix protein 74; c-SRC; F-SRC-1; Hin-2 protein; KAT13A; mNRC-1; NCOA1; NCoA-1; Nuclear receptor coactivator 1; nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; Src1; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1
Gene Symbols NCOA1
Host Species Mouse
Immunogen Synthetic peptide corresponding to residues 477-947 of human SRC-1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17977, 8648
Target Species Human, Mouse, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less