missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SRBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556506 Supplier Invitrogen™ Supplier No. PA556506
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: VWGGTCKKEE NDDDFTFGQS ALKKIKTETY PQGQPVKFPA NANSTKEEVE MNWDMVQVLS ERTNIEPWVC ANIIRLFNDD NTIPF Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

SRBD1 gene ontology annotations related to this gene include nucleobase-containing compound metabolic process; translation.

Specifications

Antigen SRBD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SRBD1
Gene Accession No. Q8N5C6
Gene Alias AI461933; C85414; D530025C17Rik; H_NH0576F01.1; RGD1562788; S1 RNA binding domain 1; S1 RNA-binding domain-containing protein 1; Srbd1; WUGSC:H_NH0576F01.1
Gene Symbols SRBD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SRBD1. Recombinant protein control fragment (Product #RP-95974).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55133
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less