missing translation for 'onlineSavingsMsg'
Learn More

Squaric Acid Dibutyl Ester, 97%

Catalog No. p-3737387
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC133170010 1 g Glass bottle
AC133170050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC133170010 Supplier Thermo Scientific Chemicals Supplier No. 133170010
Only null left

CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2892-62-8
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00037150
InChI Key XBRWELTXMQSEIN-UHFFFAOYSA-N
Synonym dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
PubChem CID 65108
ChEBI CHEBI:53612
IUPAC Name 3,4-dibutoxycyclobut-3-ene-1,2-dione
SMILES CCCCOC1=C(OCCCC)C(=O)C1=O

Specifications

Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 121.0°C to 122.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950
Quantity 1 g
Specific Gravity 1.05
Merck Index 15, 8897
Solubility Information Solubility in water: insoluble.
Formula Weight 226.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Squaric acid dibutyl ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.

RUO – Research Use Only