missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Square HDPE Reagent Bottles with Wide Mouth Polypropylene Caps
Description
- Square reagent bottles - Wide mouth square HDPE reagent bottles with polypropylene caps, designed for easy handling and efficient storage in laboratory environments.
- Leak-proof tested - Rigorously tested to ensure secure sealing under pressure, preventing spills during storage or transport.
- Chemical resistance for versatile use - Made from HDPE, these bottles are ideal for storing a wide range of chemicals and reagents.
- Laboratory and industrial applications - Perfect for chemical storage, liquid handling, and mixing in laboratories, research, and industrial settings.
- Translucent and durable design - Lightweight, translucent construction provides visibility of contents, while the square shape optimizes storage space.
- Designed for chemical storage and transport, these leak-proof, chemical-resistant bottles are ideal for laboratory and industrial applications. Non-sterile, -100°C to 120°C.
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Capacity (English) | 17 oz. |
| Color | Translucent |
| Material | HDPE |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Product Type | Reagent Bottle |