missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Squamous Epithelium

Catalog No. S30208
Change view
Click to view available options
Media Type:
Formalin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
S30208 Formalin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30208 Supplier Eisco™ Supplier No. BS18098

Prepared microscope slide of mammalian squamous epithelium. Squamous epithelium is one of epithelial tissue types. Explore structure-function relationships as per NGSS standards.

  • Pre-cleaned glass slide with ground edges
  • Professionally stained samples for best visualization
  • Ideal for for biology classrooms to explore structure-function relationships as per NGSS standards
  • Shows characteristic structures

Specifications

Biosafety Level 1
Concentration 10%
Color Clear
Material Glass
Type Microbiology slide
Manufacturing Origin China
Media Type Formalin
Packaging Cardboard Mailer
Species Mammal
Size 1 x 3 in.
Slide Type Prepared Microbiology Slide
Show More Show Less