missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalane, NF, 97-102%, Spectrum™ Chemical
S1987, 111-01-3, C30H62
$415.12 - $24,483.52
Chemical Identifiers
| CAS | 111-01-3 |
|---|---|
| Molecular Formula | C30H62 |
| Molecular Weight (g/mol) | 422.83 |
| InChI Key | PRAKJMSDJKAYCZ-UHFFFAOYNA-N |
| IUPAC Name | 2,6,10,15,19,23-hexamethyltetracosane |
| SMILES | CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606981
|
N/A | 100 mL | Amber Glass Bottle |
Each for $415.12
|
|
||||
|
18606982
|
N/A | 500 mL | Amber Glass Bottle |
Each for $1,325.21
|
|
||||
|
18606983
|
N/A | 20 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606984
|
N/A | 12 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Squalane, NF is used as an emollient and moisturizer. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 111-01-3 | |
| 422.83 | |
| 2,6,10,15,19,23-hexamethyltetracosane |
| C30H62 | |
| PRAKJMSDJKAYCZ-UHFFFAOYNA-N | |
| CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C |
Specifications
| 111-01-3 | |
| 212°C | |
| C30H62 | |
| 2 | |
| PRAKJMSDJKAYCZ-UHFFFAOYNA-N | |
| 2,6,10,15,19,23-hexamethyltetracosane | |
| 97 to 102% | |
| Liquid |
| 1 | |
| 0.005 | |
| 1.4510-1.4525 | |
| 0.807-0.810 | |
| CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C | |
| 422.83 | |
| NF |