missing translation for 'onlineSavingsMsg'
Learn More

Squalane, NF, 97-102%, Spectrum™ Chemical

Catalog No. 18606981
Change view
Click to view available options
Quantity:
100 mL
500 mL
12 kg
20 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606981 100 mL Amber Glass Bottle
18606982 500 mL Amber Glass Bottle
18606983 20 L Amber Glass Bottle
18606984 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606981 Supplier Spectrum Chemical Mfg Cor Supplier No. S1987100ML
Get Up to $500 in Chemicals with Purchase
View all promotions

S1987, 111-01-3, C30H62

Spectrum™ Chemical Squalane, NF is used as an emollient and moisturizer. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111-01-3
Molecular Formula C30H62
Molecular Weight (g/mol) 422.83
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYNA-N
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

CAS Max % 1
Boiling Point 212°C
Ignition Residue 0.005
Packaging Amber Glass Bottle
Refractive Index 1.4510-1.4525
Quantity 100 mL
Saponification Value 2
Specific Gravity 0.807-0.810
Percent Purity 97 to 102%
Grade NF
Physical Form Liquid
Show More Show Less