missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Squalane NF
Description
Squalane NF is a colorless, practically odorless transparent oil.
Specifications
Specifications
| Chemical Name or Material | Squalane NF |
| Packaging | Bottle, Amber Glass, Boston Round, 500mL, 38-439 |
| CAS | 111-01-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 422.82 |
| Quantity | 500 mL |
| Percent Purity | 97 to 102 % |
| Grade | NF |