missing translation for 'onlineSavingsMsg'
Learn More

Medisca Squalane NF

Catalog No. 14186454
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14186454 1 L
14185385 500 mL
14185399 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14186454 Supplier Medisca Supplier No. 026009

Squalane NF is a colorless, practically odorless transparent oil.

Squalane NF is a colorless, practically odorless transparent oil.

Specifications

Chemical Name or Material Squalane NF
Packaging Bottle, 1000mL, Amber Glass, Boston Round, 33-430
CAS 111-01-3
Type Drug Standard
Molecular Weight (g/mol) 422.82
Quantity 1 L
Percent Purity 97 to 102 %
Grade NF