Learn More
Squalane, ≥88%, Thermo Scientific Chemicals
Chemical Identifiers
| CAS | 111-01-3 |
|---|---|
| Molecular Formula | C30H62 |
| Molecular Weight (g/mol) | 422.826 |
| MDL Number | MFCD00008953 |
| InChI Key | PRAKJMSDJKAYCZ-UHFFFAOYSA-N |
| Synonym | squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane |
| PubChem CID | 8089 |
| IUPAC Name | 2,6,10,15,19,23-hexamethyltetracosane |
| SMILES | CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAA1793122
|
N/A | 100 g |
N/A
|
|
|||||
|
AAA1793136
|
N/A | 500 g |
N/A
|
|
|||||
Description
Squalane influence on the vital pathway that transforms glucose into cholesterol may be pivotal. It regulates or controls the rate of synthesis of the enzyme HMG Co-A reductase, and may contribute to effective treatments for cancer and heart disease. It plays an important role as a terminator in the biomembrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityNot miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| C30H62 | |
| MFCD00008953 | |
| squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
| 2,6,10,15,19,23-hexamethyltetracosane |
Specifications
| 111-01-3 | |
| 0.81 | |
| 218°C (424°F) | |
| 1.451 | |
| 776019 | |
| squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane | |
| PRAKJMSDJKAYCZ-UHFFFAOYSA-N | |
| 2,6,10,15,19,23-hexamethyltetracosane | |
| 8089 | |
| ≥88% |
| -38°C | |
| 210°C to 215°C (1 mmHg) | |
| C30H62 | |
| MFCD00008953 | |
| 14,8767 | |
| Not miscible or difficult to mix in water. | |
| CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C | |
| 422.826 | |
| 422.83 | |
| Squalane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 203-825-6
RTECSNumber : XB6070000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only