missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Squalane, 90%, Thermo Scientific Chemicals
Description
Squalane influence on the vital pathway that transforms glucose into cholesterol may be pivotal. It regulates or controls the rate of synthesis of the enzyme HMG Co-A reductase, and may contribute to effective treatments for cancer and heart disease. It plays an important role as a terminator in the biomembrane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | -38°C |
| Density | 0.81 |
| Boiling Point | 210°C to 215°C (1 mmHg) |
| Flash Point | 218°C (424°F) |
| Refractive Index | 1.451 |
| Quantity | 100 g |
| Beilstein | 776019 |
| Merck Index | 14,8767 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 422.83 |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 203-825-6 |
| RTECSNumber | XB6070000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only