missing translation for 'onlineSavingsMsg'
Learn More

SPTAN1/Alpha II-spectrin Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

Specifications

Antigen SPTAN1/Alpha II-spectrin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
501559882
Encompass
View Documents Promotion Details
N/A 20 μL N/A
Only null left
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

Spectrins are a family of filamentous cytoskeletal proteins that function as essential scaffold proteins that stabilize the plasma membrane and organize intracellular organelles. Spectrins are composed of alpha and beta dimers that associate to form tetramers linked in a head-to-head arrangement. This gene encodes an alpha spectrin that is specifically expressed in nonerythrocytic cells. The encoded protein has been implicated in other cellular functions including DNA repair and cell cycle regulation. Mutations in this gene are the cause of early infantile epileptic encephalopathy-5. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

SPTAN1/Alpha II-spectrin
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
(ALPHA)II-SPECTRIN, FLJ44613, NEAS
Between 1000 and 1050
Primary
4° C
SPTAN1
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse
P16546, Q13813
20740, 6709
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents