missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPOP Recombinant Rabbit Monoclonal Antibody (23GB4650)
GREENER_CHOICE

Catalog No. MA552722
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552722 100 μL
MA552723 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552722 Supplier Invitrogen™ Supplier No. MA552722
Only null left

Rabbit Recombinant Monoclonal Antibody

SPOP Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Component of a cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex that mediates the ubiquitination of target proteins, leading most often to their proteasomal degradation.

Specifications

Antigen SPOP
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4650
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SPOP
Gene Accession No. O43791
Gene Alias AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140
Gene Symbols SPOP
Host Species Rabbit
Immunogen A synthesized peptide derived from human SPOP (300-335AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8405
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less