missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Specifications
| For Use With (Application) | SPME |
|---|---|
| Type | SPME Fibers and Accessories |
| Material | Fused Silica Fiber |
| Quantity | 3/Pk. |
| Catalog Number | Mfr. No. | For Use With (Equipment) | Needle Gauge | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | For Use With (Equipment) | Needle Gauge | Price | Quantity & Availability | ||||
|
111020749
|
MilliporeSigma™ Supelco™
57343U |
Autosampler | 23 ga |
N/A
|
|
||||
|
111020753
|
MilliporeSigma™ Supelco™
57319 |
Autosampler | 24 ga |
N/A
|
|
||||
|
111008485
|
MilliporeSigma™ Supelco™
57344U |
Manual Holder | 23 ga |
N/A
|
|
||||
|
111008581
|
MilliporeSigma™ Supelco™
57318 |
Manual Holder | 24 ga |
N/A
|
|
||||
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.Specifications
| SPME | |
| SPME Fibers and Accessories | |
| Fused Silica Fiber | |
| 3/Pk. |