missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Supplier: MilliporeSigma™ Supelco™ 57344U
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.Chemical Identifiers
| SPME | |
| SPME Fibers and Accessories | |
| Fused Silica Fiber | |
| 3/Pk. |
Specifications
| SPME | |
| SPME Fibers and Accessories | |
| 23 ga |
| Manual Holder | |
| Fused Silica Fiber | |
| 3/Pk. |