missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Spironolactone, 99%

$117.67 - $381.51

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.57
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
Synonym spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC207460010
View Documents Promotion Details
Thermo Scientific™
207460010
1g Glass bottle
Each for $117.67
Only null left
 
AC207460050
View Documents Promotion Details
Thermo Scientific™
207460050
5g Glass bottle
Each for $381.51
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

52-01-7
416.57
LXMSZDCAJNLERA-ZHYRCANASA-N
5833
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
C24H32O4S
MFCD00082250
spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
CHEBI:9241
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

52-01-7
Authentic
C24H32O4S
15, 8887
− 37.00
Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C
416.57
CHEBI:9241
99%
0.5% max. (105°C, 2 hrs)
98.5% min. (HPLC)
MFCD00082250
−37° (20°C c=1,CHCl3)
spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
LXMSZDCAJNLERA-ZHYRCANASA-N
S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
5833
416.57
Spironolactone, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 200-133-6

RTECSNumber : TU4725000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only