missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Spironolactone, 99%

Catalog No. AC207460050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207460050 5 g Glass bottle
AC207460010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207460050 Supplier Thermo Scientific Chemicals Supplier No. 207460050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52-01-7 | C24H32O4S | 416.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52-01-7
Molecular Formula C24H32O4S
Molecular Weight (g/mol) 416.57
MDL Number MFCD00082250
InChI Key LXMSZDCAJNLERA-ZHYRCANASA-N
Synonym spironolactone, aldactone, spirolactone, verospiron, euteberol, spiroctan, spirolang, verospirone, aldactone a, spironocompren
PubChem CID 5833
ChEBI CHEBI:9241
IUPAC Name S-[(7R,8R,9S,10R,13S,14S,17R)-10,13-dimethyl-3,5'-dioxospiro[2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17,2'-oxolane]-7-yl] ethanethioate
SMILES CC(=O)SC1CC2=CC(=O)CCC2(C3C1C4CCC5(C4(CC3)C)CCC(=O)O5)C

Specifications

Melting Point 198.0°C to 207.0°C
Color White to White-Yellow
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 8887
Specific Rotation Condition −37° (20°C c=1,CHCl3)
Specific Rotation − 37.00
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 416.57
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Spironolactone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-133-6
RTECSNumber TU4725000
TSCA TSCA

RUO – Research Use Only