missing translation for 'onlineSavingsMsg'
Learn More

Spirobicromane 99.0+%, TCI America™

Catalog No. S03835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03835G Supplier TCI America Supplier No. S03835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3127-14-8
Molecular Formula C21H24O4
Molecular Weight (g/mol) 340.419
MDL Number MFCD00145320
InChI Key BWQOXWHQZXVXSD-UHFFFAOYSA-N
Synonym 7,7′C-Dihydroxy-4,4,4′C,4′C-tetramethyl-2,2′C-spirobichroman, 4,4,4′C,4′C-Tetramethyl-2,2′C-spirobi[chroman]-7,7′C-diol
PubChem CID 592148
IUPAC Name 4,4,4',4'-tetramethyl-2,2'-spirobi[3H-chromene]-7,7'-diol
SMILES CC1(CC2(CC(C3=C(O2)C=C(C=C3)O)(C)C)OC4=C1C=CC(=C4)O)C

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 340.42
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Spirobicromane