missing translation for 'onlineSavingsMsg'
Learn More

Spiramycin 90.0+%, TCI America™

Supplier:  TCI America S057525G

Quantity 25 g

 View more versions of this product

Catalog No. S057525G


Only null left

Description

Description

Specifications

Chemical Identifiers

8025-81-8
843.065
ACTOXUHEUCPTEW-ZOTSFZJCSA-N
131674974
CC1CC=CC=CC(C(CC(C(C(C(CC(=O)O1)O)OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)OC4CCC(C(O4)C)N(C)C
C43H74N2O14
MFCD01314545
Formacidine
2-[(4R,5S,6S,7R,9R,10R,16R)-6-[(2S,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-10-[(2R,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxa

Specifications

Spiramycin
Crystalline Powder
151°C
25 g
MFCD01314545
ACTOXUHEUCPTEW-ZOTSFZJCSA-N
2-[(4R,5S,6S,7R,9R,10R,16R)-6-[(2S,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-10-[(2R,5S,6R)-5-(dimethylamino)-6-methyloxan-2-yl]oxy-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxa
131674974
≥90.0% (HPLC)
Spiramycin
8025-81-8
White/Yellow
C43H74N2O14
Formacidine
CC1CC=CC=CC(C(CC(C(C(C(CC(=O)O1)O)OC)OC2C(C(C(C(O2)C)OC3CC(C(C(O3)C)O)(C)O)N(C)C)O)CC=O)C)OC4CCC(C(O4)C)N(C)C
843.065
843.07
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0468&(9)-2294

RTECSNumber : WG9400000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions