missing translation for 'onlineSavingsMsg'
Learn More

Spiramycin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015586
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015586 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015586 Supplier Merck Emd Millipore Supplier No. PHR1950500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Spiramycin belongs to the macrolide group of antibiotics, widely used in the treatment of respiratory tract, skin and skin-structure, sexually transmitted, and various other infections. Spiramycin is derived from the culture filtrate of Streptomyces ambofaciens and its mode of action involves the exibition of antimicrobial activity by inhibiting ribosomal protein biosynthesis.

Specifications

CAS 8025-81-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
MDL Number MFCD01314545
Synonym Spiramycin; Formacidine
Type Certified Reference Material
Quantity 500 mg
Grade Certified Reference Material