missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SPINeasy™ Virus RNA Kit
Description
- SPINeasy Virus RNA Kit is a silica membrane spin column kit that enables quick and convenient extraction of viral RNA from cell culture media and bodily fluids, such as saliva and serum, without the need for phenol or chloroform extractions.
- Through a simple workflow, viral RNA is typically extracted within 20 minutes and is immediately available for downstream applications. including RT-PCR, qPCR, and RNA-Seq.
Specifications
Specifications
| Content And Storage | Store at Room Temperature, Dry Environment |
| Format | Spin Column |
| Isolation Technology | Spin Column |
| Sample Type | Cell Culture Media and Bodily Fluids (Serum and Saliva); 100 to 500 µL of sample |
| Includes | 30 mL Lysis Buffer VR, 6 mL Wash Buffer VR, 10 mL Nuclease-free water, 1 vial Carrier RNA, 50 x Column VR with collection tubes |
| Quantity | 50 Preps |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.