missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals SPINeasy™ RNA Kit for Yeast

Catalog No. MP116565050
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116565050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116565050 Supplier MP Biomedicals Supplier No. 116565050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A high-performance RNA extraction kit designed for efficient purification using silica membrane spin-column technology.

SPINeasy™ RNA Kit for Yeast is a high-performance RNA extraction kit which is developed based on silica-membrane spin-column technology. By combining with beads beating method which could homogenize the yeast cell walls within 40 sec, the SPINeasy™ RNA Kit for Yeast is capable of isolating RNA from a wide range of yeast species.

  • Enables reliable RNA isolation across a broad range of yeast species.
  • When used in combination with a bead-beating approach, yeast cell walls can be effectively homogenized in as little as 40 sec, supporting consistent and efficient RNA recovery.

Specifications

Buffer Lysis Buffer
Control Type RNA Control
Isolation Technology Silica Spin Column
Species All
Sample Type Yeast
No. of Reactions 50 Preps
Product Type RNA Kit for Yeast
Sterility Non-sterile
Target Total RNA