missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals SPINeasy™ RNA Kit for Urine
Description
- Produces high-integrity RNA without phenol or chloroform, immediately ready for real-time PCR, RT-PCR, and sequencing applications.
- RNA MATRIX is a proprietary silica-based binding matrix offering flexibility in RNA binding capacities and selectively binds RNA for purification.
- The RNA is preferentially purified from other cellular components such as proteins, without the use of phenol or chloroform.
- The resulting RNA is of high integrity and suitable for various downstream applications, including real-time PCR, reverse transcription PCR (RT-PCR), and sequencing.
Specifications
Specifications
| Buffer | Lysis Buffer |
| Control Type | RNA Control |
| Isolation Technology | Spin Column |
| Species | All |
| Sample Type | Urine |
| No. of Reactions | 50 Preps |
| Product Type | RNA Kit for Urine |
| Sterility | Non-sterile |
| Target | Total RNA |