missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals SPINeasy™ RNA Kit for Feces

Catalog No. MP116556050
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116556050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116556050 Supplier MP Biomedicals Supplier No. 116556050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Delivers high yields of pure RNA in a fast and efficient workflow.

  • Provides pure RNA without detectable DNA contamination in under 20 minutes.
  • Optimized for downstream applications such as qPCR and RT-qPCR, eliminating the need for additional inhibitor or DNA removal steps.

Specifications

Buffer Lysis Buffer
Control Type RNA Control
Isolation Technology Silica Spin Column
Species All
Sample Type Feces
No. of Reactions 50 Preps
Product Type RNA Kit for Feces
Sterility Non-sterile
Target Total RNA