missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals SPINeasy™ RNA Kit for Blood

Catalog No. MP116566050
Change view
Click to view available options
No. of Reactions:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116566050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116566050 Supplier MP Biomedicals Supplier No. 116566050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Engineered to deliver superior RNA quality through a fast and simplified purification workflow.

  • Advanced RNA extraction solution based on state-of-the-art silica membrane spin-column technology, ensuring superior yield, purity, and integrity.
  • Supports a rapid and efficient workflow with fast on-column DNase treatment, enabling effective removal of co-purified DNA in as little as a few minutes.
  • Designed for short overall processing times, allowing complete RNA purification in under a quarter of an hour.
  • Streamlines blood RNA isolation by eliminating the need for erythrocyte lysis and leukocyte centrifugation steps.
  • Incorporates a simplified buffer system consisting of a single lysis buffer and two optimized wash buffers, effectively removing salts, metabolites, and large cellular components while minimizing handling complexity.

Specifications

Buffer Lysis Buffer
Control Type RNA Control
Isolation Technology Silica Spin Column
Species All
Sample Type Blood
No. of Reactions 50 Preps
Product Type RNA Kit for Blood
Target Total RNA