missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ RNA Kit for Bacteria

Catalog No. MP116541050
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116541050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116541050 Supplier MP Biomedicals™ Supplier No. 116541050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Quick and convenient purification of total RNA from microbial samples using spin column technology. Lysing Matrix B and specially formulated Lysis Buffer for efficient and reproducible sample lysis with FastPrep instruments.

  • SPINeasy RNA Kit for Bacteria is a silica -membrane spin column kit that enables quick and convenient purification of total RNA from microbial samples, including bacteria and fungi.
  • Included in the kit is our specially formulated RNASS solution that stabilizes and protects RNA.
  • The use of Lysis Buffer R and Lysing Matrix B in combination with FastPrep instruments from MP Biomedicals enables highly efficient lysis of microbial samples within seconds. With a simple workflow, the kit allows multiple samples to be processed simultaneously, without the use of toxic substances such as phenol and chloroform.
  • Total RNA of high quality and integrity can be purified typically within 40 minutes and is immediately available for RT-PCR and other downstream applications.

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Spin Column
Isolation Technology Spin Column
Sample Type Bacteria, Fungi, Yeast
Includes 50 x Lysing Matrix B, 15 mL RNASS, 60 mL Lysis Buffer R, 6 mL Wash Buffer R, 10 mL Nuclease-free water, 1 vial DNase I, 5 mL DNase I Buffer, 50 x Column R with collection tube
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.