missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ PCR Purification and Gel Extraction Kit

Catalog No. MP116538050
Change view
Click to view available options
No. of Reactions:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
MP116538050 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116538050 Supplier MP Biomedicals™ Supplier No. 116538050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Spin column kit for quick and convenient DNA clean up from PCR and other enzymatic reactions. Isolate and purify DNA fragments from agarose gels using spin column technology. Purified DNA, free of primers, enzymes, nucleotides, agarose, etc., is ready for downstream molecular biology applications.

  • The SPINeasy PCR Purification and Gel Extraction Kit is a silica -membrane spin column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gels.
  • Up to 23 µg of DNA of molecular weight ranging from 100 bp to 20 kb can be purified through a quick and simple process from most grades of agarose gels with expected recovery of 85%.
  • Purified DNA is immediately ready for routine molecular biology applications.

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Includes 50 mL Binding Buffer PG, 6 mL Wash Buffer PG, 10 mL Elution Buffer PG, 50 x Column PG with collection tubes
No. of Reactions 50

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.