missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113319
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113319 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113319 Supplier Invitrogen™ Supplier No. PA5113319

Rabbit Polyclonal Antibody

SPIN2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Human SPIN2 expressed by 2 IL3-dependent murine myeloid cell lines was moderately antiapoptotic following IL3 removal, but it was unable to block apoptosis induced by the chemotherapy agent doxorubicin. On the contrary, SPIN2-expressing cells were more sensitive to doxorubicin-mediated cell death. Also, SPIN2 did not combat the proapoptotic effects of Fas ligand stimulation in the Jurkat human T-cell line. Deletion of the C-terminal amino acids of SPIN2 diminished its antiapoptotic activity. In the murine myeloid cells, SPIN2 overexpression increased the cell number doubling times and slowed their rate of growth. There was also an increased percentage of cells in G2/M, which was more pronounced following IL3 withdrawal.

Specifications

Antigen SPIN2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SPIN2B
Gene Accession No. Q6NVE3, Q9BPZ2
Gene Alias BC068162; dJ323P24.2; Spin2; SPIN-2; SPIN2_duplicate; SPIN2B; SPIN-2B; Spin2c; SPIN-2C; Spindlin 2B; spindlin family member 2B; spindlin family, member 2; spindlin family, member 2 duplicate; spindlin family, member 2B; spindlin family, member 2C; spindlin-2; spindlin-2B; Spindlin-2C; spindlin-like protein 2; spindlin-like protein 2B; Spindlin-like protein 2C; TDRD26; telomeric SPIN2 copy
Gene Symbols SPIN2B, Spin2c
Host Species Rabbit
Immunogen Recombinant Human Spindlin-2B protein (1-258AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 278240, 474343
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less