missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Spill Response Plus (SRP) Universal Plus Absorbents

Catalog No. 19140740
Change view
Click to view available options
Dimensions (L x W):
10 x 7.5 in.
Type:
Sorbent Pad
Packaging Quantity:
100/Pk.
200/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Type Packaging Quantity
19140740 10 x 7.5 in. Sorbent Pad 100/Pk.
19140739 10 x 7.5 in. Sorbent Pad 200/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140740 Supplier Brady™ Supplier No. SRPH100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Small custom size reduces waste

  • For preventing slips, trips and falls that commonly occur in the retail, hospitality and grocery markets
  • Dimpled design for added durability and reduced linting
  • Highly absorbent, multi-purpose absorbent made to handle virtually any liquid
  • Available as pads and rolls
  • Specifications

    Material Polypropylene
    Type Sorbent Pad
    Packaging Quantity 100/Pk.
    Weight Capacity Heavyweight
    Dimensions (L x W) 10 x 7.5 in.
    Length (English) 10 in.
    Width (English) 7.5 in.