Learn More
FyterTech Nonwovens Spilfyter™ Universal Pads and Rolls
Universal sorbents for acids or bases, and nonaggressive spills, such as oil, coolants, solvents, and water
Supplier:  FyterTech Nonwovens G91
 View more versions of this product
Catalog No. 19066581
Only null left
Description
Description
Sorbents are manufactured with eight layers of high-quality, fine fiber polyethylene. Premium sorbents are bonded every 4 in., Economy every 8 in.
  • For better fit in irregular spaces, rolls are perforated horizontally every 12 in.and vertically down the center of the roll
  • For more flexible fit in narrow spaces, pads are perforated vertically
  • Premium pads and rolls offer extended use and high durability
  • Economy pads and rolls are ideal for one-time applications
  • Use pads around machinery, to line shelves or for cleanups
  • Use rolls in assembly and production areas
  • Denim Blue color inconspicuously hides dirt and grime for MRO applications
Specifications
Chemical Identifiers
OSHA 29 CFR 1910
Polyethylene Fiber
Acids or bases, and non-aggressive spills, as oil, coolants, solvents and water
Specifications
OSHA 29 CFR 1910
64 gal.
Polyethylene Fiber
Acids or bases, and non-aggressive spills, as oil, coolants, solvents and water
150 ft. x 16 in.
45.7 m
40.6 cm
15 kg
Premium Green
242.2 L
Universal Sorbent Roll
Double-weight
150 ft.
16 in.
32 lb.
Product Suggestions
Product Suggestions