missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FyterTech Nonwovens Spilfyter™ Mercury Spill Kit Powders
Refills for Spilfyter Mercury Spill Kit
$72.83 - $174.69
Specifications
| Format | Powder |
|---|---|
| Weight (English) | 1 lb. |
| Weight (Metric) | 0.45 kg |
Alert:
Chemicals do not alter the mercury's toxicity or its danger. Recommended for use by trained personnel only.
Description
Designed to handle all stages of mercury spill cleanup, quickly and safelyl, up to 0.75 oz. (25mL,) and prevent toxic vapor contamination.
- Mercury indicator powder identifies mercury in area of suspected contamination
- After an undisturbed period, powder changes color to indicate contact with mercury metal or vapor
- Mercsorb™ powder converts mercury into an amalgam for safe, convenient cleanup of mercury on work surfaces, floors and hard-to-reach areas
Specifications
| Powder | |
| 1 lb. | |
| 0.45 kg |