Learn More
FyterTech Nonwovens Spilfyter™ Lab and Cleanroom Kollect-a-Kem™ Sorbents
Available in low-lint and extra-low-lint pads and rolls
Supplier:  FyterTech Nonwovens JV222016
 View more versions of this product
Catalog No. 19-021856
Only null left
Explore available promotions
Description
Description
Low-Lint Pads and Pillows
  • Use in work areas to keep them safe from spills and chemical overruns
  • Constructed from premium fine fiber Spilfyter Hazmat Pads encased in tear-resistant, Low-Lint Spunbond fabric
Extra-Low-Lint Pads and Rolls
  • Made from Spilfyter Premium Hazmat Sorbents with extra-low-lint heat-pressed material
  • Provide for a 100% lint-free cleanroom
  • Note: It is the end user's responsibility to determine if these sorbents are suitable for use prior to allowing them into the cleanroom environment.
Specifications
Specifications
Yellow
71.9 L
Low-lint Sorbent Pillow
Heavyweight
18 in.
18 in.
16 lb.
19 gal.
Polypropylene
Keep work areas safe, free from spills and from liquid or chemical over-run
18 x 18 in. (45.72 x 45.72 cm)
45.72 cm
45.72 cm
7 kg
Product Suggestions
Product Suggestions