missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spike Sample Analysis Standard 2 (Soil) For ICP/MS, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC CLSPIKE2
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ba | 250μg/mL |
| Cr | 250μg/mL |
| Cu | 250μg/mL |
| Zn | 250μg/mL |
| V | 150μg/mL |
| Ni | 125μg/mL |
| Co | 100μg/mL |
| Pb | 100μg/mL |
| Sb | 100μg/mL |
| As | 50μg/mL |
| Cd | 50μg/mL |
| Ag | 25μg/mL |
| Be | 25μg/mL |
| Se | 25μg/mL |
| Tl | 25μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Spike Sample 2 | |
| Triple-leached LDPE bottle | |
| GC-MS | |
| Certified Reference Material | |
| 125 mL | |
| Components in 5% HNO3/tr. Tartaric Acid/tr. HF: Sb •100μg/mL As •50μg/mL Ba •250μg/mL Be •25μg/mL Cd •50μg/mL Cr •250μg/mL Co •100μg/mL Cu •250μg/mL Pb •100μg/mL Ni •125μg/mL Se •25μg/mL Ag •25μg/mL Tl •25μg/mL V •150μg/mL Zn •250μg/mL |
| For Soil | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride | |
| ICP/ICP-MS |
Product intended for laboratory use only