missing translation for 'onlineSavingsMsg'
Learn More

SPI1 Mouse anti-Human, Clone:5C7, Abnova™

Catalog No. 89334548
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89334548 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89334548 Supplier Abnova Corporation Supplier No. MAB17735
Only null left

Mouse monoclonal antibody raised against recombinant human SPI1.

This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Antigen SPI1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5C7
Conjugate Unconjugated
Description spleen focus forming virus (SFFV) proviral integration oncogene spi1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene SPI1
Gene Alias OF, PU.1, SFPI1, SPI-1, SPI-A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 124-271 of human SPI1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 6688
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less