missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPHK2 Mouse anti-Human, Clone:9C5E1, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human SPHK2.
Supplier: Abnova Corporation MAB17143
Specifications
| SPHK2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot Immunohistochemistry (1:200-1:1000) Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| SPHK2 | |
| A synthetic peptide corresponding to amino acid 36-52 of human SPHK2. | |
| 56848 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 9C5E1 | |
| sphingosine kinase 2 | |
| In PBS (0.05% sodium azide) | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG1 |