missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPHK1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302177AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Sphingosine Kinase (SphK) catalyzes the phosphorylation of the lipid sphingosine, creating the bioactive lipid sphingosine-1-phosphate (S1P). S1P subsequently signals through cell surface G protein-coupled receptors, as well as intracellularly, to modulate cell proliferation, survival, motility and differentiation. SphK is an important signaling enzyme which is activated by diverse agents, including growth factors that signal through receptor tyrosine kinases, agents activating G protein-coupled receptors, and immunoglobulin receptors. Two SphK isotypes, SphK-1 and SphK-2, have been cloned, and both isotypes are ubiquitously expressed. SphK-1 has been shown to mediate cell growth, prevention of apoptosis, and cellular transformation, and is upregulated in a variety of human tumors. In contrast, SphK-2 increases apoptosis, and may be responsible for phosphorylating and activating the immunosuppressive drug FTY720.Specifications
| SPHK1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9NYA1 | |
| SPHK1 | |
| Between 334 and 384 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SPHK1 | |
| RP23-193A16.4, 1110006G24Rik, Sk1, Spk1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8877 | |
| 4° C | |
| Liquid |