accessibility menu, dialog, popup

UserName

H3C(R)OH(S)NH2OPOHOHO(E)

CAS RN 26993-30-6

IUPAC Name: {[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
Synonyms: sphingosine 1-phosphate sphingosine-1-phosphate sphing-4-enine 1-phosphate sphingosine 1-phosphic acid S1P compound C18-sphingosine 1-phosphate
Molecular Weight (g/mol): 379.48
Molecular Formula: C18H38NO5P
InChi Key: DUYSYHSSBDVJSM-KRWOKUGFSA-N
SMILES: CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
Product Type 
  • (2)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results