Learn More
Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Pellets
Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with these Pellets.
Supplier:  Cole-Parmer™ 3623C
 View more versions of this product
Catalog No. 3623C
Only null left
Description
Description
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
  • Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
  • The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.
Specifications
Chemical Identifiers
Pellet
Specifications
Pellet
31 mm
For 3623 31 mm Evacuable Pellet Die Set
1.22 in.
For Routine Pressing of Abrasive Materials
Tungsten
Product Suggestions
Product Suggestions