Learn More
Cole-Parmer™ Hydraulic Laboratory Pellet Press Accessories, Pellets
Optimize performance of your Cole-Parmer™ hydraulic laboratory pellet press with these Pellets.
Supplier:  Cole-Parmer™ 3614C
 View more versions of this product
Catalog No. 3614C
Only null left
Description
Description
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more.
  • Ensure top results from your Cole-Parmer™ hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components.
  • The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.
Specifications
Chemical Identifiers
Pellet
Specifications
Pellet
40 mm
For 3614 40 mm Evacuable Pellet Die Set
1.5 in.
For Routine Pressing of Non-Abrasive Materials
Tungsten
Product Suggestions
Product Suggestions