missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Grinding Vial Set, Small Vial, PC Cylinder and End Plugs, PC Encapsulated Steel Impactor

Catalog No. 6761
Change view
Click to view available options
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
6761 Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6761 Supplier Cole-Parmer™ Supplier No. 6761

Small Grinding Vial grinds soft plant and animal tissues and eliminates metal contamination.

  • Small Poly-Vial Set contains everything necessary to make one complete grinding vial plus three spare polycarbonate center cylinders.
  • Grinds one sample at a time in 6775 Freezer/Mill.
  • Grinds four samples at a time in Large Freezer/Mill when used in conjunction with 6806 Multi-Vial Adapter.
  • Sample grinding capacity: 0.5 to 4 g (0.5 to 4.0 mL)
  • Withstands liquid nitrogen temperatures.
  • Closed grinding vial prevents sample cross-contamination, retains volatiles, and controls hazardous or critical samples.
  • Transparent plastic center cylinder allows visual checking of the grinding progress.
  • Disposable polycarbonate center cylinders.
  • Polycarbonate end plugs, impactor, and center cylinders may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Compatible with 6754 Small Extractor/Vial Opener.

Order Info

Replacement impactors and end plugs are available (See Cat. Nos. 6761E and 6761P).

Specifications

Product Type Grinding Set
Capacity (English) 0.02 to 0.14 oz.
Capacity (Metric) 0.5 to 4 mL
For Use With (Application) Ideal for Grinding Soft Plant and Animal Tissues and Eliminates Metal Contamination
For Use With (Equipment) Small Extractor/Vial Opener and Auto Extractor
Includes 1 Small Polycarbonate Encapsulated Impactor, 2 Small Polycarbonate End Plugs, and 1 Small Polycarbonate Center Cylinder
Material Polycarbonate
Sample Volume (English) 0.01 to 0.13 oz.
Sample Volume (Metric) 0.5 to 4 mL
Size Small
Weight (English) 0.017 to 0.17 oz.
Weight (Metric) 0.5 to 5 g
Show More Show Less