Learn More
Cole-Parmer™ Replacement Cylinders for Mid-Size Grinding Vial Sets
Replacement Cylinders for 6881 mid-size grinding vial set, 6883 mid-size low-chrome grinding vial set, and 6885 Mid-size poly-vial set.
Supplier:  Cole-Parmer™ 6881C4
Catalog No. 68-81C-4
Only null left
Description
Description
  • Withstands liquid nitrogen temperatures.
  • Are disposable or may be cleaned with hot water and detergent or RNase/DNase-free solution.
  • Transparent plastic center cylinder allows visual checking of the grinding progress.
Specifications
Specifications
Center Cylinder
Mid Size, Cr-Free, Poly-Vial Grinding Vial Set
Polycarbonate
4 Pk.
Product Suggestions
Product Suggestions