Learn More
SPEX™ SamplePrep Large-Capacity Freezer/Mill™ Vial Divider for Grinding Vial Set 6751
Catalog No. 6807
Change view
Click to view available options
Holds:
Microvial set
Catalog No. Holds
6807 Microvial set
1 options
Catalog No. 6807 Supplier SPEX™ SamplePrep Supplier No. 6807
Only null left
For use with SPEX SamplePrep™ Large-Capacity Freezer/Mill Cryogenic Grinder
  • Makes one complete vial plus three spare center sections
  • Sample capacity: 0.5 to 4.0mL
  • Includes stainless-steel impactor, two end plugs
Specifications
Product Type Multi Vial Adapter
For Use With (Application) Ideal for high throughput applications
For Use With (Equipment) Spex SamplePrep Large Capacity Freezer/Mill
Holds Microvial set